missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-thien-2-yl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3175752
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
871825-56-8
C9H8N2O2S
208.24
MFCD08271954
YSTOGMCNCZGBTG-UHFFFAOYSA-N
1-methyl-3-thien-2-yl-1h-pyrazole-5-carboxylic acid, 1-methyl-3-thiophen-2-yl-1h-pyrazole-5-carboxylic acid, 2-methyl-5-thiophen-2-yl-2h-pyrazole-3-carboxylic acid, 2-methyl-5-thiophen-2-yl pyrazole-3-carboxylic acid, 1-methyl-3-2-thienyl-1 h-pyrazole-5-carboxylic acid, 1-methyl-3-2-thienyl pyrazole-5-carboxylic acid, 2-methyl-5-2-thienyl pyrazole-3-carboxylic acid, 5-carboxy-1-methyl-3-thien-2-yl-1h-pyrazole, 2-5-carboxy-1-methyl-1h-pyrazol-3-yl thiophene
20109897
2-methyl-5-thiophen-2-ylpyrazole-3-carboxylic acid
CN1N=C(C=C1C(O)=O)C1=CC=CS1
This item is not returnable. View return policy
871825-56-8
C9H8N2O2S
208.24
MFCD08271954
YSTOGMCNCZGBTG-UHFFFAOYSA-N
1-methyl-3-thien-2-yl-1h-pyrazole-5-carboxylic acid, 1-methyl-3-thiophen-2-yl-1h-pyrazole-5-carboxylic acid, 2-methyl-5-thiophen-2-yl-2h-pyrazole-3-carboxylic acid, 2-methyl-5-thiophen-2-yl pyrazole-3-carboxylic acid, 1-methyl-3-2-thienyl-1 h-pyrazole-5-carboxylic acid, 1-methyl-3-2-thienyl pyrazole-5-carboxylic acid, 2-methyl-5-2-thienyl pyrazole-3-carboxylic acid, 5-carboxy-1-methyl-3-thien-2-yl-1h-pyrazole, 2-5-carboxy-1-methyl-1h-pyrazol-3-yl thiophene
20109897
2-methyl-5-thiophen-2-ylpyrazole-3-carboxylic acid
CN1N=C(C=C1C(O)=O)C1=CC=CS1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 208.24
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-thien-2-yl-1H-pyrazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'