missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-pyrrolidinol, 97%, Thermo Scientific Chemicals

Catalog Number 10527844
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13220-33-2
C5H11NO
101.15
MFCD00003176
FLVFPAIGVBQGET-UHFFFAOYSA-N
1-methyl-3-pyrrolidinol, 3-hydroxy-1-methylpyrrolidine, n-methyl-3-pyrrolidinol, 3-pyrrolidinol, 1-methyl, 3-hydroxy-n-methylpyrrolidine, n-methyl-3-hydroxypyrrolidine, 1-methyl-3-hydroxypyrrolidine, pubchem14117, l-methyl-3-pyrrolidinol, 1-methyl-pyrrolidin-3ol
93074
1-methylpyrrolidin-3-ol
CN1CCC(C1)O
This item is not returnable. View return policy
13220-33-2
C5H11NO
101.15
MFCD00003176
FLVFPAIGVBQGET-UHFFFAOYSA-N
1-methyl-3-pyrrolidinol, 3-hydroxy-1-methylpyrrolidine, n-methyl-3-pyrrolidinol, 3-pyrrolidinol, 1-methyl, 3-hydroxy-n-methylpyrrolidine, n-methyl-3-hydroxypyrrolidine, 1-methyl-3-hydroxypyrrolidine, pubchem14117, l-methyl-3-pyrrolidinol, 1-methyl-pyrrolidin-3ol
93074
1-methylpyrrolidin-3-ol
CN1CCC(C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Density 0.9210g/mL
Boiling Point 50.0°C to 52.0°C (1.0 mmHg)
Flash Point 71°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, II, 3
Specific Gravity 0.921
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-pyrrolidinol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methyl-3-Pyrrolidinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'