missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Methyl-3-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63787.03

Code : NEW

Additional Details : CAS Number : 18391-77-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15460669

  • £49.82 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

1-Methyl-3-phenyl-2-thiohydantoin
206.26
168827
18391-77-0
MFCD00464059
1-Methyl-3-phenyl-2-thioxo-4-imidazolidinone

Specifications

1-Methyl-3-phenyl-2-thiohydantoin
18391-77-0
95%
MFCD00464059
1-Methyl-3-phenyl-2-thioxo-4-imidazolidinone
1g
Solid
206.26
168827
Videos
SDS