missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-octylimidazolium chloride, 97%, Acros Organics

Brand:  Acros Organics 356610050

UNSPSC : 12161500

Code : 29

Additional Details : CAS Number : 64697-40-1 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11906261

  • £23.80 / 5g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

1-Methyl-3-octylimidazolium chloride
97%
C12H23ClN2
230.78

Specifications

1-Methyl-3-octylimidazolium chloride
64697-40-1
97%
1.0124
230.78
5g
1.5060 to 1.5080
97%
C12H23ClN2
1.0124g/mL
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Methyl-3-octylimidazolium chloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents