missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-(2-thienyl)-1H-pyrazol-5-amine, 97%, Maybridge

Brand:  Maybridge KM00835EA

Code : 31

Additional Details : CAS Number : 118430-78-7 Weight : 0.06000kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10232642

  • £176.00 / 10g


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Specifications

Chemical Identifiers

1-Methyl-3-(2-thienyl)-1H-pyrazol-5-amine
95.0
95%
179.25
118430-78-7
95%
C8H9N3S
MFCD00068075

Specifications

1-Methyl-3-(2-thienyl)-1H-pyrazol-5-amine
118430-78-7
Amber Glass Bottle
95%
179.25
10g
95.0
95%
C8H9N3S
MFCD00068075
Videos
SDS