missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-imidazolidinone, 98+%, Thermo Scientific Chemicals

Catalog Number p-163915
Click to view available options
Quantity:
500 mg
2 g
Packaging:
Glass bottle
This item is not returnable. View return policy
694-32-6
C4H8N2O
100.12
MFCD00800601
JTPZTKBRUCILQD-UHFFFAOYSA-N
567600
1-methylimidazolidin-2-one
CN1CCNC1=O
This item is not returnable. View return policy
694-32-6
C4H8N2O
100.12
MFCD00800601
JTPZTKBRUCILQD-UHFFFAOYSA-N
567600
1-methylimidazolidin-2-one
CN1CCNC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 500 mg
Synonym 1-methyl-2-imidazolidinone, 2-imidazolidinone, 1-methyl, 1-methyl-2-imidazolidone, n-methylimidazolidinone, acmc-209yb9, 3-methylimidazolidin-2-one, 1-methylimidazolidine-2-one, 1-methyl-imidazolidin-2-one, 2-imidazolidinone,1-methyl, 1-methyl-2-imidazolidinone #
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 100.12
Percent Purity 98+%
Physical Form Crystalline Solid
Chemical Name or Material 1-Methyl-2-imidazolidinone, 98+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'