missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-3-carbonitrile, 97%, Maybridge

Brand:  Maybridge CC48316DE

Code : 31

Additional Details : CAS Number : 79080-39-0 Weight : 0.05500kg

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10515364

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

1-Methyl-1H-pyrazole-3-carbonitrile
96.5
97%
C5H5N3
MFCD00466343
79080-39-0
100.0
97%
107.12

Specifications

1-Methyl-1H-pyrazole-3-carbonitrile
79080-39-0
100.0
97%
C5H5N3
MFCD00466343
5g
96.5
Amber Glass Bottle
97%
107.12
Videos
SDS