missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-1h-pyrazole-3-carbaldehyde, 97%, Maybridge

Brand:  Maybridge MO07706DE

Code : 31

Additional Details : CAS Number : 27258-32-8 Weight : 0.05500kg

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10718825

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

1-Methyl-1H-pyrazole-3-carbaldehyde
97%
110.12
97%
C5H6N2O

Specifications

1-Methyl-1H-pyrazole-3-carbaldehyde
27258-32-8
97%
C5H6N2O
5g
Amber Glass Bottle
97%
110.12
Videos
SDS