missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazol-5-ylamine, 97%, Thermo Scientific™

Catalog Number p-2695265
Click to view available options
Quantity:
1 g
10 g
100 g
This item is not returnable. View return policy
1192-21-8
C4H7N3
97.121
MFCD00068156
JESRNIJXVIFVOV-UHFFFAOYSA-N
1-methyl-1h-pyrazol-5-amine, 1-methyl-1h-pyrazol-5-ylamine, 1-methyl-5-aminopyrazole, 5-amino-1-methylpyrazole, 5-amino-1-methyl-1h-pyrazole, 1h-pyrazol-5-amine, 1-methyl, 2-methyl-2h-pyrazol-3-ylamine, 5-amino-1-methyl-pyrazole, 2-methyl-2h-pyrazol-3-amine, 1-methylpyrazole-5-ylamine
136927
2-methylpyrazol-3-amine
CN1C(=CC=N1)N
This item is not returnable. View return policy
1192-21-8
C4H7N3
97.121
MFCD00068156
JESRNIJXVIFVOV-UHFFFAOYSA-N
1-methyl-1h-pyrazol-5-amine, 1-methyl-1h-pyrazol-5-ylamine, 1-methyl-5-aminopyrazole, 5-amino-1-methylpyrazole, 5-amino-1-methyl-1h-pyrazole, 1h-pyrazol-5-amine, 1-methyl, 2-methyl-2h-pyrazol-3-ylamine, 5-amino-1-methyl-pyrazole, 2-methyl-2h-pyrazol-3-amine, 1-methylpyrazole-5-ylamine
136927
2-methylpyrazol-3-amine
CN1C(=CC=N1)N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 97.12
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazol-5-ylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'