missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrazol-3-yl)methylamine, 97%, Maybridge

Brand:  Maybridge CC48313CB

Code : 31

Additional Details : CAS Number : 612511-81-6 Weight : 0.05025kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10451134

  • £47.00 / 250mg


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Chemical Identifiers

(1-Methyl-1H-pyrazol-3-yl)methylamine
96.5
97%
C5H9N3
MFCD03419807
612511-81-6
100.0
95%
111.15

Specifications

(1-Methyl-1H-pyrazol-3-yl)methylamine
612511-81-6
100.0
97%
C5H9N3
MFCD03419807
250mg
96.5
Amber Glass Bottle
95%
111.15
Videos
SDS