Learn More
1-Methyl-1H-indole-5-sulfonyl chloride, 90%, Maybridge
Brand:  Maybridge CC41403CB
Code : 50
Additional Details : CAS Number : 859850-75-2 Weight : 0.05025kg Transport : UN number : 3261 Chem class : 8 Pack group : III
Quantity 250mg
Packaging Amber Glass Bottle
 View more versions of this product
Product Code. 10476263
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications
Chemical Identifiers
1-Methyl-1H-indole-5-sulfonyl chloride
89.5
90%
C9H8ClNO2S
MFCD08271907
859850-75-2
100.0
90%
229.69
Specifications
1-Methyl-1H-indole-5-sulfonyl chloride
89.5
Amber Glass Bottle
90%
229.69
250mg
859850-75-2
100.0
90%
C9H8ClNO2S
MFCD08271907