1-methyl-1h-indole-4-carboxylic acid, 95%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC45801DA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 90924-06-4 Weight: 0.05100kg Transport: UN number : 2811 Chem class : 1 Pack group : III


Packaging Amber Glass Bottle
Quantity 1g

Product Code. 10202703

Quantity Price
1 £ 185.0 / 1g
Estimated Shipment date
from Supplier 03-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-indole-4-carboxylic acid
CAS 90924-06-4
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD08435874

Specification

Formula Weight 175.19
Quantity 1g
CAS Min % 94.5
Assay 95%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 90924-06-4
MDL Number MFCD08435874
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Packaging Amber Glass Bottle
Molecular Formula C10H9NO2
Health Hazard 1 GHS Signal Word: Warning
CAS Max % 100.0
Chemical Name or Material 1-Methyl-1H-indole-4-carboxylic acid
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.