(1-Methyl-1H-indol-7-yl)methylamine, 90%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC66913DA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 937795-97-6 Weight: 0.00100kg Transport: UN number : 2735 Chem class : 1b Pack group : III


Packaging Amber Glass Bottle
Quantity 1g

Product Code. 10612895

Quantity Price
1 £ 183.0 / 1g
Estimated Shipment date
from Supplier 04-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material (1-Methyl-1H-indol-7-yl)methylamine
CAS 937795-97-6
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C10H12N2
Formula Weight 160.22
MDL Number MFCD09879961

Specification

Formula Weight 160.22
Quantity 1g
Assay 90%
CAS Min % 89.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 937795-97-6
MDL Number MFCD09879961
Assay Percent Range 90%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Packaging Amber Glass Bottle
Molecular Formula C10H12N2
Health Hazard 1 GHS Signal Word: Danger
CAS Max % 100.0
Chemical Name or Material (1-Methyl-1H-indol-7-yl)methylamine
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.