missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indol-5-amine, 97%, Thermo Scientific™

Catalog Number p-3175471
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
102308-97-4
C9H10N2
146.19
MFCD03839873
PGTSGPCXPIFQEL-UHFFFAOYSA-N
1-methyl-1h-indol-5-amine, 5-amino-1-n-methylindole, 1-methyl-1h-indol-5-ylamine, 1h-indol-5-amine, 1-methyl, 5-amino-1-methylindole, 1-methylindole-5-ylamine, 1-methyl-5-amino-1h-indole, 5-amino-1-methyl-1h-indole, pubchem9381, 5-amino-n-methylindole
2769564
1-methylindol-5-amine
CN1C=CC2=CC(N)=CC=C12
This item is not returnable. View return policy
102308-97-4
C9H10N2
146.19
MFCD03839873
PGTSGPCXPIFQEL-UHFFFAOYSA-N
1-methyl-1h-indol-5-amine, 5-amino-1-n-methylindole, 1-methyl-1h-indol-5-ylamine, 1h-indol-5-amine, 1-methyl, 5-amino-1-methylindole, 1-methylindole-5-ylamine, 1-methyl-5-amino-1h-indole, 5-amino-1-methyl-1h-indole, pubchem9381, 5-amino-n-methylindole
2769564
1-methylindol-5-amine
CN1C=CC2=CC(N)=CC=C12
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indol-5-amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'