missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-indazol-3-yl)methylamine dihydrochloride, 97%, Thermo Scientific™

Catalog Number p-3881263
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1093860-45-7
C9H13Cl2N3
234.124
MFCD11841065
ZHWICCCSNYFLJY-UHFFFAOYSA-N
1-methyl-1h-indazol-3-yl methylamine dihydrochloride, 1-methylindazol-3-yl methanamine dihydrochloride, 1-methyl-1h-indazol-3-yl methanamine dihydrochloride, 3-aminomethyl-1-methyl-1h-indazole dihydrochloride, 1-methyl-1h-indazol-3-yl methylamine 2hcl, 1-methyl-1h-indazol-3-yl methylamine, chloride, chloride, 1-1-methylindazol-3-yl methanamine dihydrochloride, 1-1-methyl-1h-indazol-3-yl methanamine-hydrogen chloride 1/2
43811028
(1-methylindazol-3-yl)methanamine;dihydrochloride
CN1C2=CC=CC=C2C(=N1)CN.Cl.Cl
This item is not returnable. View return policy
1093860-45-7
C9H13Cl2N3
234.124
MFCD11841065
ZHWICCCSNYFLJY-UHFFFAOYSA-N
1-methyl-1h-indazol-3-yl methylamine dihydrochloride, 1-methylindazol-3-yl methanamine dihydrochloride, 1-methyl-1h-indazol-3-yl methanamine dihydrochloride, 3-aminomethyl-1-methyl-1h-indazole dihydrochloride, 1-methyl-1h-indazol-3-yl methylamine 2hcl, 1-methyl-1h-indazol-3-yl methylamine, chloride, chloride, 1-1-methylindazol-3-yl methanamine dihydrochloride, 1-1-methyl-1h-indazol-3-yl methanamine-hydrogen chloride 1/2
43811028
(1-methylindazol-3-yl)methanamine;dihydrochloride
CN1C2=CC=CC=C2C(=N1)CN.Cl.Cl
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 234.13
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-indazol-3-yl)methylamine dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'