1-Methyl-1H-imidazole-5-carbonitrile, 97+%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC23616CB

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 66121-66-2 Weight: 0.05025kg Transport: UN number : 3439 Chem class : 1 Pack group : III


Packaging Amber Glass Bottle
Quantity 250mg

Product Code. 10576713

Quantity Price
1 £ 44.0 / 250mg
Estimated Shipment date
from Supplier 01-06-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-imidazole-5-carbonitrile
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5N3
Formula Weight 107.12

Specification

Formula Weight 107.12
Quantity 250mg
Assay 97%
Chemical Name or Material 1-Methyl-1H-imidazole-5-carbonitrile
CAS 66121-66-2
Packaging Amber Glass Bottle
Molecular Formula C5H5N3
Assay Percent Range 97%
Documentation

Technical documents