missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-4-sulfonamide, 97%, Thermo Scientific™

Catalog Number p-2695849
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
111124-90-4
C4H7N3O2S
161.18
MFCD02091382
QBJSSOBNVYDCDQ-UHFFFAOYSA-N
2736885
1-methylimidazole-4-sulfonamide
CN1C=NC(=C1)S(N)(=O)=O
This item is not returnable. View return policy
111124-90-4
C4H7N3O2S
161.18
MFCD02091382
QBJSSOBNVYDCDQ-UHFFFAOYSA-N
2736885
1-methylimidazole-4-sulfonamide
CN1C=NC(=C1)S(N)(=O)=O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-imidazole-4-sulfonamide, 1h-imidazole-4-sulfonamide,1-methyl, 1-methyl-1h-imidazole-4-sulphonamide, acmc-20dtvv, maybridge1_000050, spr_7, 1-methyl-4-imidazolesulfonamide, 1-methyl-1h-imidazol-4-ylsulfonamide, 1h-imidazole-4-sulfonamide, 1-methyl
Formula Weight 161.18
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-imidazole-4-sulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'