missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-imidazole-4-carboxylic acid, 97%, Maybridge
Click to view available options
Amber Glass Bottle; 250mg
Specifications
CAS | 41716-18-1 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C5H6N2O2 |
Quantity | 250 mg |
Synonym | 1-methyl-1h-imidazole-4-carboxylic acid, 1-methyl-imidazole-4-carboxylic acid, 1-methyl-4-imidazole-carboxylic acid, 1-methyl-4-imidazolecarboxylicacid, pubchem9001, acmc-1am6x, 1-methyl-4-imidazolic acid, ksc237q3j, 1-methyl-4-imidazolecarboxylic acid, 1-methyl-1h-4carboxylic acid |
InChI Key | WZTRQGJMMHMFGH-UHFFFAOYSA-N |
SMILES | CN1C=C(N=C1)C(=O)O |
IUPAC Name | 1-methylimidazole-4-carboxylic acid |
Molecular Weight (g/mol) | 126.115 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'