missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-2-carboxylic acid, Maybridge

Brand:  Maybridge CC20801DA

Code : 31

Additional Details : CAS Number : 20485-43-2 Weight : 0.05100kg

Quantity 1g
Packaging Amber glass bottle

 View more versions of this product

Product Code. 10160772

  • £48.41 / 1g


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Specifications

Chemical Identifiers

1-Methyl-1H-imidazole-2-carboxylic acid
97%
126.11
97%
C5H6N2O2

Specifications

1-Methyl-1H-imidazole-2-carboxylic acid
Amber glass bottle
97%
126.11
20485-43-2
97%
C5H6N2O2
1g
Videos
SDS