missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Methyl-1H-imidazol-5-yl)methylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 111.15 |
Percent Purity | 97% |
Chemical Name or Material | (1-Methyl-1H-imidazol-5-yl)methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'