missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-imidazol-4-yl)methylamine, 95%, Thermo Scientific™

Catalog Number p-4509860
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
486414-83-9
C5H9N3
111.15
MFCD06411536
YSEAGFBRAQOCFM-UHFFFAOYSA-N
2795114
(1-methylimidazol-4-yl)methanamine
CN1C=NC(CN)=C1
This item is not returnable. View return policy
486414-83-9
C5H9N3
111.15
MFCD06411536
YSEAGFBRAQOCFM-UHFFFAOYSA-N
2795114
(1-methylimidazol-4-yl)methanamine
CN1C=NC(CN)=C1
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methyl-1h-imidazol-4-yl methanamine, 1-methyl-1h-imidazol-4-yl methylamine, 1h-imidazole-4-methanamine, 1-methyl, 1-methylimidazol-4-yl methanamine, 4-aminomethyl-1-methylimidazole, c-1-methyl-1h-imidazol-4-yl-methylamine, 1-1-methylimidazol-4-yl methanamine, 4-aminomethyl-1-methyl-1h-imidazole, 1h-imidazole-4-methanamine,1-methyl, 1-methyl-1h-imidazol-4-yl methyl amine
Formula Weight 111.15
Percent Purity 95%
Chemical Name or Material (1-Methyl-1H-imidazol-4-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'