missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Methyl-1H-imidazol-4-yl)methylamine, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 1-methyl-1h-imidazol-4-yl methanamine, 1-methyl-1h-imidazol-4-yl methylamine, 1h-imidazole-4-methanamine, 1-methyl, 1-methylimidazol-4-yl methanamine, 4-aminomethyl-1-methylimidazole, c-1-methyl-1h-imidazol-4-yl-methylamine, 1-1-methylimidazol-4-yl methanamine, 4-aminomethyl-1-methyl-1h-imidazole, 1h-imidazole-4-methanamine,1-methyl, 1-methyl-1h-imidazol-4-yl methyl amine |
Formula Weight | 111.15 |
Percent Purity | 95% |
Chemical Name or Material | (1-Methyl-1H-imidazol-4-yl)methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'