missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-imidazol-4-yl)methanol, 97%, Maybridge

Brand:  Maybridge CC03609DA

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 17289-25-7 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10026332

  • £165.00 / 1g


Estimated Shipment date
from Supplier 01-06-2017
Add to basket


Specifications Description & Specifications

Chemical Identifiers

(1-Methyl-1H-imidazol-4-yl)methanol
96.5
97%
C5H8N2O
MFCD06738905
17289-25-7
100.0
97%
112.13

Specifications

112.13
97%
17289-25-7
97%
C5H8N2O
(1-Methyl-1H-imidazol-4-yl)methanol
1g
96.5
MFCD06738905
Amber Glass Bottle
100.0
Documentation

Technical Documents