missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-benzimidazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3586605
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
53484-17-6
C9H8N2O2
176.175
MFCD06809665
KAJLCBKTWRUQOI-UHFFFAOYSA-N
1-methyl-1h-benzimidazole-5-carboxylic acid, 1-methyl-1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-1,3-benzodiazole-5-carboxylic acid, 1-methyl-1,3-benzodiazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1-methyl, 1-methyl-5-benzimidazolecarboxylic acid, 1-methyl-1h-benzimidazole-5-carboxylicacid, 1-methylbenzimidazol-5-carboxylic acid, 1-methylbenzimidazole-5-carboxylicacid
4913063
1-methylbenzimidazole-5-carboxylic acid
CN1C=NC2=C1C=CC(=C2)C(=O)O
This item is not returnable. View return policy
53484-17-6
C9H8N2O2
176.175
MFCD06809665
KAJLCBKTWRUQOI-UHFFFAOYSA-N
1-methyl-1h-benzimidazole-5-carboxylic acid, 1-methyl-1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylic acid, 1-methyl, 1-methyl-1h-1,3-benzodiazole-5-carboxylic acid, 1-methyl-1,3-benzodiazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1-methyl, 1-methyl-5-benzimidazolecarboxylic acid, 1-methyl-1h-benzimidazole-5-carboxylicacid, 1-methylbenzimidazol-5-carboxylic acid, 1-methylbenzimidazole-5-carboxylicacid
4913063
1-methylbenzimidazole-5-carboxylic acid
CN1C=NC2=C1C=CC(=C2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-benzimidazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'