missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1-cyclohexene, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number p-161672
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
591-49-1
CTMHWPIWNRWQEG-UHFFFAOYSA-N
1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
11574
1-methylcyclohexene
CC1=CCCCC1
This item is not returnable. View return policy
591-49-1
CTMHWPIWNRWQEG-UHFFFAOYSA-N
1-methyl-1-cyclohexene, cyclohexene, 1-methyl, 1-methylcyclohex-1-ene, cyclohexene, methyl, methylcyclohexene, .alpha.-methylcyclohexene, 1-methyl-cyclohexene, methyl-1-cyclohexene, 2,3,4,5-tetrahydrotoluene, unii-te4p8q2044
11574
1-methylcyclohexene
CC1=CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Density 0.8100g/mL
Boiling Point 110.0°C to 111.0°C
Flash Point −3°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12
Linear Formula C6H9CH3
Refractive Index 1.4492 to 1.4512
Quantity 10 g
Beilstein 05, 66
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,acetone and benzene
Stabilizer 10ppm hydroquinone
Molecular Weight (g/mol) 96.17
Formula Weight 96.17
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 1-Methyl-1-cyclohexene, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methyl-1-cyclohexene, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'