missing translation for 'onlineSavingsMsg'
Learn More

1-Mesitylhydrazine hydrochloride, 97%, ACROS Organics™

Brand:  Acros Organics 438340050

Code : 29

Additional Details : CAS Number : 24006-09-5 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 11905671

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

1-Mesitylhydrazine hydrochloride
96.0
97%
C9H14N2·HCl
24006-09-5
100.0
97%
186.69

Specifications

1-Mesitylhydrazine hydrochloride
96.0
Authentic
Powder or Crystalline Powder
97%
186.69
175.0° to 178.5°C
24006-09-5
100.0
Glass bottle
97%
C9H14N2·HCl
White to tan
5g
Videos
SDS