missing translation for 'onlineSavingsMsg'
Learn More

1-(Mesitylenesulfonyl)imidazole, 98+%, Thermo Scientific™

Catalog Number p-7022332
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
50257-39-1
C12H14N2O2S
250.32
MFCD00005284
XFHVQSPNDDOABS-UHFFFAOYSA-N
1-mesitylsulfonyl-1h-imidazole, 1-2-mesitylenesulfonyl imidazole, n-mesitylenesulfonyl imidazole, n-mesitylenesulfonylimidazole, 1-2,4,6-trimethylbenzenesulfonyl imidazole, 1-2,4,6-trimethylphenyl sulphonyl-1h-imidazole, 1h-imidazole, 1-2,4,6-trimethylphenyl sulfonyl, 2,4,6-trimethylphenyl sulfonyl imidazole, acmc-209kkw, mesitylenesulfonylimidazole
99556
1-(2,4,6-trimethylphenyl)sulfonylimidazole
CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=CN=C1
This item is not returnable. View return policy
50257-39-1
C12H14N2O2S
250.32
MFCD00005284
XFHVQSPNDDOABS-UHFFFAOYSA-N
1-mesitylsulfonyl-1h-imidazole, 1-2-mesitylenesulfonyl imidazole, n-mesitylenesulfonyl imidazole, n-mesitylenesulfonylimidazole, 1-2,4,6-trimethylbenzenesulfonyl imidazole, 1-2,4,6-trimethylphenyl sulphonyl-1h-imidazole, 1h-imidazole, 1-2,4,6-trimethylphenyl sulfonyl, 2,4,6-trimethylphenyl sulfonyl imidazole, acmc-209kkw, mesitylenesulfonylimidazole
99556
1-(2,4,6-trimethylphenyl)sulfonylimidazole
CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=CN=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 97°C to 99°C
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 250.32
Percent Purity ≥98%
Chemical Name or Material 1-(Mesitylenesulfonyl)imidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'