missing translation for 'onlineSavingsMsg'
Learn More

1-(Mercaptomethyl)cyclopropaneacetic acid, 98%, Thermo Scientific™

Catalog Number 15438288
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
162515-68-6
C6H10O2S
146.204
MFCD03411600
VFAXPOVKNPTBTM-UHFFFAOYSA-N
2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid
9793825
2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
C1CC1(CC(=O)O)CS
This item is not returnable. View return policy
162515-68-6
C6H10O2S
146.204
MFCD03411600
VFAXPOVKNPTBTM-UHFFFAOYSA-N
2-1-mercaptomethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropaneacetic acid, 1-mercaptomethyl cyclopropylacetic acid, 1-mercaptomethyl-cyclopropaneacetic acid, unii-1x00t93wyx, cyclopropaneacetic acid, 1-mercaptomethyl, 2-1-sulfanylmethyl cyclopropyl acetic acid, 1-mercaptomethyl cyclopropyl acetic acid, 1-sulfanylmethyl cyclopropyl acetic acid
9793825
2-[1-(sulfanylmethyl)cyclopropyl]acetic acid
C1CC1(CC(=O)O)CS

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 42°C to 46°C
Color White
Flash Point >110°C (230°F)
Odor Stench
Quantity 1 g
UN Number UN3261
Formula Weight 146.21
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-(Mercaptomethyl)cyclopropaneacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'