missing translation for 'onlineSavingsMsg'
Learn More

1-Isoquinolinecarbonitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-4812617
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1198-30-7
C10H6N2
154.17
HJHXYSBRTVFEDD-UHFFFAOYSA-N
1-cyanoisoquinoline, 1-isoquinolinecarbonitrile, isoquinaldonitrile, isoquinolinecarbonitrile, 1-cyano-isoquinoline, isoquinaldinic nitrile, pubchem6269, maybridge1_002114, timtec-bb sbb002487
306057
isoquinoline-1-carbonitrile
C1=CC=C2C(=C1)C=CN=C2C#N
This item is not returnable. View return policy
1198-30-7
C10H6N2
154.17
HJHXYSBRTVFEDD-UHFFFAOYSA-N
1-cyanoisoquinoline, 1-isoquinolinecarbonitrile, isoquinaldonitrile, isoquinolinecarbonitrile, 1-cyano-isoquinoline, isoquinaldinic nitrile, pubchem6269, maybridge1_002114, timtec-bb sbb002487
306057
isoquinoline-1-carbonitrile
C1=CC=C2C(=C1)C=CN=C2C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 90.0°C to 92.0°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 154.17
Percent Purity 98%
Chemical Name or Material 1-Isoquinolinecarbonitrile
compliance-icons
Product Identifier
  • 1-Isoquinolinecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'