missing translation for 'onlineSavingsMsg'
Learn More

1-Isopropyl-4-piperidone, 99%, Thermo Scientific Chemicals

Catalog Number p-7081480
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
5355-68-0
C8H15NO
141.214
MFCD00038035
CCDBCHAQIXKJCG-UHFFFAOYSA-N
1-isopropyl-4-piperidone, 1-isopropylpiperidin-4-one, n-isopropyl-4-piperidone, 4-piperidinone, 1-1-methylethyl, 1-isopropyl-4-piperidinone, 4-piperidone, 1-isopropyl, 1-propan-2-yl piperidin-4-one, 1-1'-methylethyl-4-piperidone, 1-methylethyl piperidin-4-one
79313
1-propan-2-ylpiperidin-4-one
CC(C)N1CCC(=O)CC1
This item is not returnable. View return policy
5355-68-0
C8H15NO
141.214
MFCD00038035
CCDBCHAQIXKJCG-UHFFFAOYSA-N
1-isopropyl-4-piperidone, 1-isopropylpiperidin-4-one, n-isopropyl-4-piperidone, 4-piperidinone, 1-1-methylethyl, 1-isopropyl-4-piperidinone, 4-piperidone, 1-isopropyl, 1-propan-2-yl piperidin-4-one, 1-1'-methylethyl-4-piperidone, 1-methylethyl piperidin-4-one
79313
1-propan-2-ylpiperidin-4-one
CC(C)N1CCC(=O)CC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.95
Boiling Point 100°C to 101°C (27 mmHg)
Refractive Index 1.4655
Quantity 10 g
Beilstein 107857
Formula Weight 141.22
Percent Purity 99%
Chemical Name or Material 1-Isopropyl-4-piperidone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'