missing translation for 'onlineSavingsMsg'
Learn More

1-Iodonaphthalene, 97.5%, Thermo Scientific Chemicals

Catalog Number p-3758234
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
90-14-2
C10H7I
254.07
MFCD00003876
NHPPIJMARIVBGU-UHFFFAOYSA-N
naphthalene, 1-iodo, 1-naphthyl iodide, alpha-iodonaphthalene, iodonaphthalene, 1-iodonaphthalenen, .alpha.-iodonaphthalene, 1-iodo-naphthalene, 1-iodonaphthlen, 1-iodonaphthlene, 1-iodonapthalene
7004
1-iodonaphthalene
C1=CC=C2C(=C1)C=CC=C2I
This item is not returnable. View return policy
90-14-2
C10H7I
254.07
MFCD00003876
NHPPIJMARIVBGU-UHFFFAOYSA-N
naphthalene, 1-iodo, 1-naphthyl iodide, alpha-iodonaphthalene, iodonaphthalene, 1-iodonaphthalenen, .alpha.-iodonaphthalene, 1-iodo-naphthalene, 1-iodonaphthlen, 1-iodonaphthlene, 1-iodonapthalene
7004
1-iodonaphthalene
C1=CC=C2C(=C1)C=CC=C2I

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.7400g/mL
Boiling Point 302.0°C to 305.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96.5% min. (GC)
Packaging Glass bottle
Linear Formula C10H7I
Refractive Index 1.6980 to 1.7040
Quantity 10 mL
Beilstein 05, 550
Specific Gravity 1.74
Formula Weight 254.07
Percent Purity 97.5%
Physical Form Liquid
Chemical Name or Material 1-Iodonaphthalene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Iodonaphthalene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'