missing translation for 'onlineSavingsMsg'
Learn More

1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3758242
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2050-77-3
C10H21I
268.18
MFCD00001085
SKIDNYUZJPMKFC-UHFFFAOYSA-N
16314
1-iododecane
CCCCCCCCCCI
This item is not returnable. View return policy
2050-77-3
C10H21I
268.18
MFCD00001085
SKIDNYUZJPMKFC-UHFFFAOYSA-N
16314
1-iododecane
CCCCCCCCCCI

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.2500g/mL
Boiling Point 124.0°C (11.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)9I
Refractive Index 1.4845 to 1.4865
Quantity 25 g
Beilstein 01,168
Specific Gravity 1.25
Synonym decyl iodide, decane, 1-iodo, n-decyl iodide, iododecane, 1-iodanyldecane, 1-decyl iodide, 1-iodo-decane, 1-iod0decane, decane,1-iodo, pubchem3942
Stabilizer copper
Formula Weight 268.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iododecane
Show More Show Less
compliance-icons
Product Identifier
  • 1-Iododecane, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'