missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2-methylpropane, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number p-149017
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
513-38-2
C4H9I
184.02
MFCD00001084
BTUGGGLMQBJCBN-UHFFFAOYSA-N
isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane
10556
1-iodo-2-methylpropane
CC(C)CI
This item is not returnable. View return policy
513-38-2
C4H9I
184.02
MFCD00001084
BTUGGGLMQBJCBN-UHFFFAOYSA-N
isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane
10556
1-iodo-2-methylpropane
CC(C)CI

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -93.0°C
Color Colorless to Orange
Density 1.5900g/mL
Boiling Point 120.0°C to 121.0°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2I
Refractive Index 1.4948 to 1.4968
Quantity 25 mL
Merck Index 15,5189
Specific Gravity 1.59
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Stabilizer copper
Formula Weight 184.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Iodo-2-methylpropane, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Iodo-2-methylpropane, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'