missing translation for 'onlineSavingsMsg'
Learn More

1-Indanone, 99+%, Thermo Scientific Chemicals

Catalog Number 10735571
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
This item is not returnable. View return policy
83-33-0
C9H8O
132.16
QNXSIUBBGPHDDE-UHFFFAOYSA-N
1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone
6735
CHEBI:17404
2,3-dihydroinden-1-one
C1CC(=O)C2=CC=CC=C21
This item is not returnable. View return policy
83-33-0
C9H8O
132.16
QNXSIUBBGPHDDE-UHFFFAOYSA-N
1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone
6735
CHEBI:17404
2,3-dihydroinden-1-one
C1CC(=O)C2=CC=CC=C21

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 38.0°C to 42.0°C
Color Brown to Yellow
Density 1.1000g/mL
Boiling Point 243.0°C to 245.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 07,36
Specific Gravity 1.1
Solubility Information Solubility in water: 6.5g/L (20°C). Other solubilities: soluble in ethanol,ether and chloroform
Formula Weight 132.16
Percent Purity 99+%
Physical Form Crystalline Mass
Chemical Name or Material 1-Indanone
Show More Show Less
compliance-icons
Product Identifier
  • 1-Indanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'