missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-^m-hydrotetracarbonyldiruthenium(II), 98%, Thermo Scientific Chemicals

Catalog Number p-7080237
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy
104439-77-2
C62H42O6Ru2
MFCD03840556
Shvo's Catalyst
This item is not returnable. View return policy
104439-77-2
C62H42O6Ru2
MFCD03840556
Shvo's Catalyst

1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-μ-hydroteracarbonyldiruthenium(II) is widely used in various types of applications in organic synthesis. It is used for hydrogenation of carbonyls, alkenes and imines. It is also used in the alkylation of amines and amination of propargyl alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Calayst for hydrogen transfers including reduction of ketones and aldehydes, and oxidation and/or racemization of amines and alcohols
TRUSTED_SUSTAINABILITY
Melting Point 223°C to 227°C
Color Yellow to Orange
Quantity 0.5 g
Sensitivity Moisture sensitive
Merck Index 14,8483
Solubility Information Soluble in water and polar organic solvents.
Formula Weight 1085.15
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-Hydroxytetraphenylcyclopentadienyl (tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldir

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'