missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxyethylidenebis(phosphonic acid), 96%, Thermo Scientific Chemicals

Catalog Number 11315847
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
2809-21-4
C2H8O7P2
206.027
MFCD00070585
DBVJJBKOTRCVKF-UHFFFAOYSA-N
etidronic acid, etidronate, hedp, 1-hydroxyethylidene-1,1-diphosphonic acid, ehdp, etidronsaeure, acetodiphosphonic acid, hydroxyethanediphosphonic acid, oxyethylidenediphosphonic acid, turpinal sl
3305
CHEBI:4907
(1-hydroxy-1-phosphonoethyl)phosphonic acid
CC(O)(P(=O)(O)O)P(=O)(O)O
This item is not returnable. View return policy
2809-21-4
C2H8O7P2
206.027
MFCD00070585
DBVJJBKOTRCVKF-UHFFFAOYSA-N
etidronic acid, etidronate, hedp, 1-hydroxyethylidene-1,1-diphosphonic acid, ehdp, etidronsaeure, acetodiphosphonic acid, hydroxyethanediphosphonic acid, oxyethylidenediphosphonic acid, turpinal sl
3305
CHEBI:4907
(1-hydroxy-1-phosphonoethyl)phosphonic acid
CC(O)(P(=O)(O)O)P(=O)(O)O

1-Hydroxyethylidenebis(phosphonic acid) acts as human protein tyrosine phosphatase inhibitor. It is also useful for photographic applications. It finds application to strengthen bone, osteoporosis treatment and to treat Paget's disease of bone. In addition, it is used in corrosion inhibition in circulating cool water systems, oil fields and low-pressure boilers with electric power, the chemical industry, metallurgy and fertilizers. It is also utilized for formulating reverse osmosis and thermal desalination anti-scaling agents. Furthermore, it is used in detergents, water treatment, cosmetics and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Odorless
Quantity 2 g
UN Number UN3261
Merck Index 14,3863
Formula Weight 206.03
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 1-Hydroxyethylidenebis (phosphonic acid)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'