missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7081998
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4404-45-9
C7H13NS
143.248
MFCD00014445
WXYAXKKXIGHXDS-UHFFFAOYSA-N
hexyl isothiocyanate, n-hexyl isothiocyanate, hexane, 1-isothiocyanato, 1-hexyl isothiocyanate, hexylisothiocyanate, isothiocyanic acid, hexyl ester, unii-2s7c3ga0z5, 1-hexylisothiocyanate, 1-isothiocyanato-hexane, hexanisothiocyanate
78120
1-isothiocyanatohexane
CCCCCCN=C=S
This item is not returnable. View return policy
4404-45-9
C7H13NS
143.248
MFCD00014445
WXYAXKKXIGHXDS-UHFFFAOYSA-N
hexyl isothiocyanate, n-hexyl isothiocyanate, hexane, 1-isothiocyanato, 1-hexyl isothiocyanate, hexylisothiocyanate, isothiocyanic acid, hexyl ester, unii-2s7c3ga0z5, 1-hexylisothiocyanate, 1-isothiocyanato-hexane, hexanisothiocyanate
78120
1-isothiocyanatohexane
CCCCCCN=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.933
Boiling Point 216°C
Flash Point 90°C (194°F)
Odor Pungent
Refractive Index 1.4915
Quantity 5 g
UN Number UN2922
Beilstein 1751837
Sensitivity Moisture sensitive
Formula Weight 143.25
Percent Purity 97%
Chemical Name or Material 1-Hexyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'