missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexanethiol, 96%, ACROS Organics™
£20.39 - £175.00
Chemical Identifiers
CAS | 111-31-9 |
---|---|
Molecular Formula | C6H14S |
Molecular Weight (g/mol) | 118.24 |
MDL Number | MFCD00004909 |
InChI Key | PMBXCGGQNSVESQ-UHFFFAOYSA-N |
Synonym | 1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan |
PubChem CID | 8106 |
IUPAC Name | hexane-1-thiol |
SMILES | CCCCCCS |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10012930
|
Acros Organics
215270050 |
5mL | Glass bottle |
£20.39
5mL |
In stock, ready to ship: 5 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10676992
|
Acros Organics
215271000 |
100mL | Glass bottle |
£55.05
100mL |
In stock, ready to ship: 6 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10046103
|
Acros Organics
215275000 |
500mL | Glass bottle |
£175.00
500mL |
Estimated Shipment 29-01-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
111-31-9 | |
118.24 | |
PMBXCGGQNSVESQ-UHFFFAOYSA-N | |
8106 | |
CCCCCCS |
C6H14S | |
MFCD00004909 | |
1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan | |
hexane-1-thiol |
Specifications
1-Hexanethiol | |
30°C | |
Glass bottle | |
0.84 | |
Colorless to Yellow | |
5mL | |
95.0 | |
95% min. (GC) | |
CH3(CH2)5SH | |
01, 408 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohols, ethyl ether, hydrocarbons | |
CCCCCCS | |
118.24 | |
118.24 | |
96% |
0.8400g/mL | |
Authentic | |
1.4470 to 1.4510 | |
150.0°C to 154.0°C | |
-81°C to 80°C | |
111-31-9 | |
100.0 | |
C6H14S | |
MFCD00004909 | |
1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan | |
PMBXCGGQNSVESQ-UHFFFAOYSA-N | |
hexane-1-thiol | |
8106 | |
Liquid |