missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanesulfonic acid, sodium salt hydrate, 98%, HPLC grade

Brand:  Acros Organics 411601000

Code : 29

Additional Details : CAS Number : 2832-45-3 Weight : 0.15000kg

 View more versions of this product

Product Code. 10628321

  • £447.30 / 100g


Estimated Shipment
21-06-2018
Add to basket

Specifications

Chemical Identifiers

1-Hexanesulfonic acid, sodium salt hydrate
C6H13NaO3S·xH2O
188.22
04, IV, 54
2832-45-3
CH3(CH2)5SO3Na·xH2O
MFCD00149549

Specifications

0.3 max. at 200nm, 0.2 max. at 210nm, 0.1 max. at 220nm, 0.075 max. at 230nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.04 max. at 254nm, 0.04 max. at 280nm, (0.25 M aq. soln.) (1 cm cell vs HPLC grade water), (0.25 M aq. soln.) (1 cm cell vs HPLC grade water) at 0nm
Clear colorless to light yellow
(0.25M aq. soln.) (1cm cell vs HPLC grade water)
Vapor Pressure: Negligible
Authentic
Liquid
98%
CH3(CH2)5SO3Na·xH2O
MFCD00149549
04, IV, 54
1-Hexanesulfonic acid, sodium salt hydrate
100g
2832-45-3
Negligible
Glass bottle
≥97.5% (on dry substance)
C6H13NaO3S·xH2O
188.22
Solubility in water: Soluble
7 to 9%
Videos
SDS