missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanesulfonic Acid Sodium Salt, HPLC Grade, J.T. Baker™

Catalog Number 15115184
304.20 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
25 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
2832-45-3
C6H13NaO3S
188.22
MFCD00007542
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
23677630
[Na+].CCCCCCS([O-])(=O)=O
This item is not returnable. View return policy
2832-45-3
C6H13NaO3S
188.22
MFCD00007542
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
23677630
[Na+].CCCCCCS([O-])(=O)=O

High purity solvents for use in analytical and preparative separations

    For Ion-Pair Chromatography of Basic Compounds
Packaging Wide Mouth Amber Glass Bottle
Quantity 25 g
Specific Gravity 1L = 0.00kg
IUPAC Name sodium;hexane-1-sulfonate
Formula Weight 188.22
Grade HPLC
Chemical Name or Material 1-Hexanesulfonic Acid Sodium Salt
compliance-icons
Product Identifier
  • 1-HEXANESULFONIC ACID SODIUM SALT
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : SODIUM 1-HEXANESULFONATE
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'