missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexane Sulfonic Acid, Sodium Salt, 98+%, Ion Pair Chromatography, Certified, Fisher Chemical™
149.61 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
74.40 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "23271" to get your promotional price.
74.40 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
1-Hexane Sulfonic Acid, Sodium Salt, 98+%, Ion Pair Chromatography, Certified, Fisher Chemical, Quantity: 100g, CAS: 2832-45-3, White, Melting Point: 300 deg.C, Molecular Formula: C6H13NaO3S, Molecular Weight: 188.217, Packaging: Plastic powder jar
Specifications
Melting Point | 300°C |
Color | White |
Physical Form | Powder/Solid |
Quantity | 25 g |
IUPAC Name | sodium hexane-1-sulfonate |
Formula Weight | 188.22g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- 1-Hexanesulfonic acid, sodium salt
- Warning
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'