missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Hexadecyl)trimethylammonium chloride, 96%, Thermo Scientific Chemicals
16.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
(1-Hexadecyl)trimethylammonium chloride is used as a topical antiseptic and surfactant. It is used in hair care formulations, cream rinse ingredient, hair conditioners and shampoos as a conditioning agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 234°C |
Color | White |
Density | 0.968 g/mL |
Odor | Odorless |
Linear Formula | CH3(CH2)15N(CH3)3Cl |
Quantity | 5 g |
UN Number | UN1759 |
Beilstein | 3657974 |
Solubility Information | Soluble in water. |
Formula Weight | 320 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'