missing translation for 'onlineSavingsMsg'
Learn More

1-Heptyn-3-ol, 98%, Thermo Scientific™

Catalog Number p-7044222
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
7383-19-9
C7H12O
112.172
MFCD00041597
SHSFXAVQBIEYMK-UHFFFAOYSA-N
1-heptyn-3-ol, n-butyl ethynyl carbinol, acmc-1biv8, 1-heptyn-3-ol, +/-
93050
hept-1-yn-3-ol
CCCCC(C#C)O
This item is not returnable. View return policy
7383-19-9
C7H12O
112.172
MFCD00041597
SHSFXAVQBIEYMK-UHFFFAOYSA-N
1-heptyn-3-ol, n-butyl ethynyl carbinol, acmc-1biv8, 1-heptyn-3-ol, +/-
93050
hept-1-yn-3-ol
CCCCC(C#C)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Heptyn-3-ol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, heavy metal powders.
TRUSTED_SUSTAINABILITY
Density 0.869
Boiling Point 161°C to 163°C
Flash Point 59°C (138°F)
Odor Recognizable
Refractive Index 1.44
Quantity 5 g
UN Number UN1987
Beilstein 1740804
Solubility Information Slightly soluble in water.
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material 1-Heptyn-3-ol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'