missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Heptanesulfonic Acid Sodium Salt, HPLC Grade, J.T. Baker™
312.55 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
25 g
Packaging:
Wide Mouth Amber Glass Bottle
Description
- For Ion-Pair Chromatography of Basic Compounds; For Laboratory,Research,or Manufacturing Use
Specifications
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 25 g |
Specific Gravity | 1L = 0.00kg |
Formula Weight | 202.26 |
Grade | HPLC |
Chemical Name or Material | 1-Heptanesulfonic Acid Sodium Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'