missing translation for 'onlineSavingsMsg'
Learn More

1-Heptadecanol

Product Code. 11355669
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11355669

Brand: Thermo Scientific Alfa Aesar L20404.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1454-85-9 | C17H36O | 256.474 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1454-85-9
Molecular Formula C17H36O
Molecular Weight (g/mol) 256.474
MDL Number MFCD00002822
InChI Key GOQYKNQRPGWPLP-UHFFFAOYSA-N
Synonym 1-heptadecanol, heptadecyl alcohol, heptadecanol, n-heptadecanol, unii-n3il85tmcx, 1-hydroxyheptadecane, alcohols, c16-18, n3il85tmcx, heptadecanol mixed primary isomers, n-heptadecanol-1
PubChem CID 15076
ChEBI CHEBI:77470
IUPAC Name heptadecan-1-ol
SMILES CCCCCCCCCCCCCCCCCO

Specifications

Melting Point 52°C to 56°C
Boiling Point 308°C to 309°C
Flash Point 155°C (311°F)
Linear Formula CH3(CH2)16OH
Quantity 25 g
Beilstein 1702195
Formula Weight 256.47
Chemical Name or Material 1-Heptadecanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.