missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-4-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-135068
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
350-46-9
C6H4FNO2
141.1
MFCD00007282
WFQDTOYDVUWQMS-UHFFFAOYSA-N
4-fluoronitrobenzene, p-fluoronitrobenzene, p-nitrofluorobenzene, benzene, 1-fluoro-4-nitro, 4-nitrofluorobenzene, 1-nitro-4-fluorobenzene, 4-fluoro-1-nitrobenzene, 1-fluoro-4-nitro-benzene, unii-a2m2fh7xhh, p-fluoro nitrobenzene
9590
1-fluoro-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])F
This item is not returnable. View return policy
350-46-9
C6H4FNO2
141.1
MFCD00007282
WFQDTOYDVUWQMS-UHFFFAOYSA-N
4-fluoronitrobenzene, p-fluoronitrobenzene, p-nitrofluorobenzene, benzene, 1-fluoro-4-nitro, 4-nitrofluorobenzene, 1-nitro-4-fluorobenzene, 4-fluoro-1-nitrobenzene, 1-fluoro-4-nitro-benzene, unii-a2m2fh7xhh, p-fluoro nitrobenzene
9590
1-fluoro-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 27°C
Color Brown to Yellow
Density 1.3300g/mL
Boiling Point 205°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4NO2
Refractive Index 1.5302 to 1.5322
Quantity 100 g
Beilstein 05, 241
Specific Gravity 1.33
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,xylene and acetone
Formula Weight 141.1
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material 1-Fluoro-4-nitrobenzene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Fluoro-4-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'