missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-7021496
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
1493-27-2
C6H4FNO2
141.101
MFCD00007048
PWKNBLFSJAVFAB-UHFFFAOYSA-N
2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro
73895
1-fluoro-2-nitrobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])F
This item is not returnable. View return policy
1493-27-2
C6H4FNO2
141.101
MFCD00007048
PWKNBLFSJAVFAB-UHFFFAOYSA-N
2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro
73895
1-fluoro-2-nitrobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])F

1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

Solubility
Immiscible in water.

Notes
Avoid excessive heat and light.
TRUSTED_SUSTAINABILITY
Melting Point °C to 8°C
Density 1.340
Boiling Point 215°C
Flash Point 94°C (201°F)
Assay Percent Range 99%
Refractive Index 1.5317
Quantity 50 g
UN Number UN2810
Beilstein 607262
Solubility Information Immiscible in water.
Formula Weight 141.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Fluoro-2-nitrobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'