1-Fluoro-2-iodobenzene, 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 204490050

5ML 1-Fluoro-2-iodobenzene, 99%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 348-52-7 Weight: 0.05952kg


Packaging Glass bottle
Quantity 5mL

Product Code. 10002650

Quantity Price
1 £ 10.7 / 5mL
Estimated Shipment date
from Supplier 12-12-2016
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-Fluoro-2-iodobenzene
CAS 348-52-7
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C6H4FI
Linear Formula FC6H4I
Formula Weight 222g/mol
MDL Number MFCD00001031

Specification

Infrared Spectrum Authentic
Packaging Glass bottle
Storage Information Product darkens on storage
Refractive Index 1.5899 to 1.5919 (20°C, 589nm)
Density 1.903g/mL
Color Clear light yellow
Quantity 5mL
Chemical Name or Material 1-Fluoro-2-iodobenzene
CAS 348-52-7
CAS Min % 98.5
CAS Max % 100.0
Physical Form Clear light yellow liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C6H4FI
Linear Formula FC6H4I
Formula Weight 222g/mol
MDL Number MFCD00001031
Beilstein 05, I, 119
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Documentation

Technical documents

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.