missing translation for 'onlineSavingsMsg'
Learn More

1-(Ethylsulfonyl)piperazine, 97%, Thermo Scientific™

Catalog Number p-7052723
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
62937-96-6
C6H14N2O2S
178.25
MFCD02093528
BIYGAOBOLDXNHM-UHFFFAOYSA-N
1-ethylsulfonyl piperazine, 1-ethylsulphonyl piperazine, 1-ethanesulfonyl piperazine, 1-ethylsulfonyl-piperazine, 1-ethanesulfonyl-piperazine, 1-ethanesulfonylpiperazine, ethylsulfonyl piperazine, ethanesulfonylpiperazine, ethanesulfonyl-piperazine
415294
1-ethylsulfonylpiperazine
CCS(=O)(=O)N1CCNCC1
This item is not returnable. View return policy
62937-96-6
C6H14N2O2S
178.25
MFCD02093528
BIYGAOBOLDXNHM-UHFFFAOYSA-N
1-ethylsulfonyl piperazine, 1-ethylsulphonyl piperazine, 1-ethanesulfonyl piperazine, 1-ethylsulfonyl-piperazine, 1-ethanesulfonyl-piperazine, 1-ethanesulfonylpiperazine, ethylsulfonyl piperazine, ethanesulfonylpiperazine, ethanesulfonyl-piperazine
415294
1-ethylsulfonylpiperazine
CCS(=O)(=O)N1CCNCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 69°C to 72°C
Color Brown to White
Quantity 1 g
Formula Weight 178.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Ethylsulfonyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'