missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(Ethylsulfonyl)piperazine, 97%

Brand:  Alfa Aesar™ H55503.06

Code : NEW

Additional Details : CAS Number : 62937-96-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15451778

  • £101.60 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(Ethylsulfonyl)piperazine
178.25
1-(Ethanesulfonyl)piperazine
62937-96-6
MFCD02093528

Specifications

1-(Ethylsulfonyl)piperazine
69° to 72°C
62937-96-6
97%
MFCD02093528
White to pale brown
5g
Solid
178.25
1-(Ethanesulfonyl)piperazine
Videos
SDS